missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,4,5-Trimethylbenzoic Acid 97.0+%, TCI America™
Supplier: TCI America T246925G
Specifications
2,4,5-Trimethylbenzoic Acid | |
151°C | |
C10H12O2 | |
25 g | |
CC1=CC(C)=C(C=C1C)C(O)=O | |
164.20 | |
164.20 | |
Crystalline Powder |
528-90-5 | |
White-Yellow | |
MFCD00075863 | |
QENJZWZWAWWESF-UHFFFAOYSA-N | |
2,4,5-trimethylbenzoic acid | |
10714 | |
≥97.0% (GC,T) |
Chemical Identifiers
528-90-5 | |
164.20 | |
QENJZWZWAWWESF-UHFFFAOYSA-N | |
2,4,5-trimethylbenzoic acid |
C10H12O2 | |
MFCD00075863 | |
10714 | |
CC1=CC(C)=C(C=C1C)C(O)=O |
Safety and Handling
RTECSNumber : DI0887000
TSCA : No