Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',4',5'-Trimethylacetophenone, 97%

Chemical Identifiers

Chemical Name or Material 2',4',5'-Trimethylacetophenone
CAS 2040-07-5
Molecular Formula C11H14O
Formula Weight 162.23
MDL Number MFCD00017244
Beilstein 2042909
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1061409

 
alfa aesar™
A1061409
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1061418

 
alfa aesar™
A1061418
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2',4',5'-Trimethylacetophenone
Boiling Point 246° to 248°C
Melting Point 10° to 11°C
Quantity 10g
CAS 2040-07-5
Assay 97%
Molecular Formula C11H14O
Formula Weight 162.23
MDL Number MFCD00017244
Beilstein 2042909