Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,5-Trimethyl-3-oxazoline, cis + trans, 99%

Chemical Identifiers

Chemical Name or Material 2,4,5-Trimethyl-3-oxazoline
CAS 22694-96-8
Name Note cis + trans
Molecular Formula C6H11NO
Formula Weight 113.16
MDL Number MFCD00053161
UN Number UN1993
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2222806

  SDS
alfa aesar™
B2222806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2222814

  SDS
alfa aesar™
B2222814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2222822

  SDS
alfa aesar™
B2222822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 113.16
Chemical Name or Material 2,4,5-Trimethyl-3-oxazoline
Name Note cis + trans
Assay 99%
Molecular Formula C6H11NO
MDL Number MFCD00053161
CAS 22694-96-8
UN Number UN1993
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
RTECS Number RQ5665000
Odor Characteristic
Boiling Point 45° to 47°C (12mmHg)
Quantity 5g

Applications
2,4,5-Trimethyl-3-oxazoline, cis + trans possesses the characteristic meat aroma hences used for flavoring application. Used as flavour enhancer in fish

Solubility
Soluble in alcohol, fixed oils. Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.

Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.