missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,5-Trimethoxybenzoic Acid, TCI America™

Manufacturer:  TCI America T26275G

 View more versions of this product

Catalog No. T26275G



Specifications

Specifications

2,4,5-Trimethoxybenzoic Acid
White-Yellow
5g
C10H12O5
asaronic acid, benzoic acid, 2,4,5-trimethoxy, asarylic acid, calamonic acid, acmc-209ket, 2,4,5-trimethoxybenzoicacid, ksc489o7d, 2,4,5-trimethoxy-benzoic acid, 2,4,5-trimethoxybenzoic acid, alpha-hydroxyphenylacetic acid; amygdalic acid; alpha-hydroxyphenylacetic acid; alpha-hydroxy-benzeneacetic acid
COC1=CC(=C(C=C1C(=O)O)OC)OC
212.201
212.20
≥98.0% (GC,T)
300°C
144°C
490-64-2
MFCD00002435
KVZUCOGWKYOPID-UHFFFAOYSA-N
2,4,5-trimethoxybenzoic acid
10276
Crystal-Powder at 20°C

Chemical Identifiers

490-64-2
212.201
KVZUCOGWKYOPID-UHFFFAOYSA-N
10276
COC1=CC(=C(C=C1C(=O)O)OC)OC
C10H12O5
MFCD00002435
asaronic acid, benzoic acid, 2,4,5-trimethoxy, asarylic acid, calamonic acid, acmc-209ket, 2,4,5-trimethoxybenzoicacid, ksc489o7d, 2,4,5-trimethoxy-benzoic acid, 2,4,5-trimethoxybenzoic acid, alpha-hydroxyphenylacetic acid; amygdalic acid; alpha-hydroxyphenylacetic acid; alpha-hydroxy-benzeneacetic acid
2,4,5-trimethoxybenzoic acid
Safety and Handling

Safety and Handling

EINECSNumber : (3)-4610

TSCA : Yes

Documents