missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,5-Trimethoxybenzaldehyde 98%, ACROS Organics™

Manufacturer:  Acros Organics 157941000

 View more versions of this product

Catalog No. AC157941000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2, 4, 5-Trimethoxybenzaldehyde
Glass bottle
111.0°C to 115.0°C
98%
97.5
98%
(CH3O)3C6H2CHO
08, 389
IAJBQAYHSQIQRE-UHFFFAOYSA-N
2,4,5-trimethoxybenzaldehyde
20525
Fine Crystalline Powder
Authentic
White to Beige
100g
4460-86-0
100.0
C10H12O4
MFCD00003312
asaraldehyde, asaronaldehyde, asarylaldehyde, azarylaldehyde, benzaldehyde, 2,4,5-trimethoxy, gazarin, 3,4,6-trimethoxybenzaldehyde, unii-ndu8j2q00d, 2,4,5-trimethoxy-benzaldehyde, ccris 1296
COC1=CC(=C(C=C1C=O)OC)OC
196.202
196.2
98%

Chemical Identifiers

4460-86-0
196.202
IAJBQAYHSQIQRE-UHFFFAOYSA-N
20525
COC1=CC(=C(C=C1C=O)OC)OC
C10H12O4
MFCD00003312
asaraldehyde, asaronaldehyde, asarylaldehyde, azarylaldehyde, benzaldehyde, 2,4,5-trimethoxy, gazarin, 3,4,6-trimethoxybenzaldehyde, unii-ndu8j2q00d, 2,4,5-trimethoxy-benzaldehyde, ccris 1296
2,4,5-trimethoxybenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 224-713-3

RTECSNumber : CU8460000

TSCA : TSCA

SDS Documents