missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,5-Trihydroxybenzaldehyde, TCI America™

Manufacturer:  TCI America T17665G

 View more versions of this product

Catalog No. T17665G



Specifications

Specifications

2,4,5-Trihydroxybenzaldehyde
5g
C7H6O4
benzaldehyde, 2,4,5-trihydroxy, acmc-1ag53, 4,5-dihydroxy-salicylaldehyde, 2,4,5-trihydroxy-benzaldehyde, benzaldehyde,2,4,5-trihydroxy, 2,4,5-trihydroxybenzaldehyde
C1=C(C(=CC(=C1O)O)O)C=O
154.121
154.12
≥98.0% (GC)
Green-Yellow
35094-87-2
MFCD00016592
WNCNWLVQSHZVKV-UHFFFAOYSA-N
2,4,5-trihydroxybenzaldehyde
643387
Crystal-Powder at 20°C

Chemical Identifiers

35094-87-2
154.121
WNCNWLVQSHZVKV-UHFFFAOYSA-N
643387
C1=C(C(=CC(=C1O)O)O)C=O
C7H6O4
MFCD00016592
benzaldehyde, 2,4,5-trihydroxy, acmc-1ag53, 4,5-dihydroxy-salicylaldehyde, 2,4,5-trihydroxy-benzaldehyde, benzaldehyde,2,4,5-trihydroxy, 2,4,5-trihydroxybenzaldehyde
2,4,5-trihydroxybenzaldehyde
Safety and Handling

Safety and Handling

TSCA : No

Documents