Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,5-Trifluorotoluene, 98%

Chemical Identifiers

Chemical Name or Material 2,4,5-Trifluorotoluene
CAS 887267-34-7
Formula Weight 146.11
MDL Number MFCD08458011
Synonym 1,2,4-Trifluoro-5-methylbenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6319003

 
alfa aesar™
H6319003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6319006

 
alfa aesar™
H6319006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 146.11
Chemical Name or Material 2,4,5-Trifluorotoluene
Physical Form Liquid
Assay 98%
MDL Number MFCD08458011
CAS 887267-34-7
Synonym 1,2,4-Trifluoro-5-methylbenzene
Color Colorless to pale yellow
Quantity 1g