Please login to your online account to display your discounted pricing

2,4,5-Trifluorophenylboronic acid, 97%, ACROS Organics™


Quantity 25g

Manufacturer: thermo fisher scientific  AC393800250

 View more versions of this product

Catalog No. AC393800250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 175.9
Quantity 25g
Assay Percent Range 97%
Molecular Formula C6H4BF3O2
Chemical Name or Material 2, 4, 5-Trifluorophenylboronic acid
CAS 247564-72-3

Specifications

Chemical Name or Material 2, 4, 5-Trifluorophenylboronic acid
Assay Percent Range 97%
Molecular Formula C6H4BF3O2
Formula Weight 175.9