Please login to your online account to display your discounted pricing

2,4,5-Trifluorophenol, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC308680050

 View more versions of this product

Catalog No. AC308680050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder or Chunks
Formula Weight 148.09
Quantity 5g
Color White
Melting Point 37° to 42°C
CAS Max % 100.0
Chemical Name or Material 2, 4, 5-Trifluorophenol
Molecular Formula C6H3F3O
Linear Formula F3C6HOH
Infrared Spectrum Authentic
Assay Percent Range 98%
MDL Number MFCD00061215
CAS 2268-16-8
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Flammable solid.
Packaging Glass bottle

Specifications

Chemical Name or Material 2, 4, 5-Trifluorophenol
CAS 2268-16-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H3F3O
Linear Formula F3C6HOH
Formula Weight 148.09
MDL Number MFCD00061215
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Flammable solid.

SDS & Safety Documents