Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,5-Trifluorophenol, 94%

Chemical Identifiers

Chemical Name or Material 2,4,5-Trifluorophenol
CAS 2268-16-8
Molecular Formula C6H3F3O
Formula Weight 148.09
MDL Number MFCD00061215
UN Number UN2921
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2039503

  SDS
alfa aesar™
B2039503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 148.09
Flash Point 55°C (131°F)
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00061215
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, FLAMMABLE, N.O.S.
Odor Phenol-like
Quantity 1g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H314-H318-H228
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
UN Number UN2921
Melting Point 39° to 42°C
Chemical Name or Material 2,4,5-Trifluorophenol
CAS 2268-16-8
Assay 94%
Molecular Formula C6H3F3O
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H314-H318-H228
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.