Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,5-Trifluorocinnamic acid, 97+%

$83.58 - $298.62

Chemical Identifiers

Chemical Name or Material 2,4,5-Trifluorocinnamic acid
CAS 247170-17-8
Molecular Formula C9H5F3O2
Formula Weight 202.13
MDL Number MFCD00061211
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2322903

 
alfa aesar™
B2322903
1g Each for $83.58

AAB2322906

 
alfa aesar™
B2322906
5g Each for $298.62
Description & Specifications

Specifications

Formula Weight 202.13
Chemical Name or Material 2,4,5-Trifluorocinnamic acid
Assay ≥97%
Molecular Formula C9H5F3O2
MDL Number MFCD00061211
CAS 247170-17-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 184° to 188°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.