missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,5-Trifluorobenzyl Alcohol, TCI America™

Manufacturer:  TCI America T27935G

 View more versions of this product

Catalog No. T27935G



Specifications

Specifications

2,4,5-Trifluorobenzyl Alcohol
White-Yellow
144284-25-3
MFCD00061206
NRXZCCOHXZFHBV-UHFFFAOYSA-N
(2,4,5-trifluorophenyl)methanol
2777035
Crystal-Lump at 20°C
204°C
5g
C7H5F3O
2,4,5-trifluorobenzyl alcohol, 2,4,5-trifluorophenyl methanol, 2,4,5-trifluorobenzylalcohol, benzenemethanol, 2,4,5-trifluoro, 2,4,5-trifluoro-phenyl-methanol, 2,4,5-trifluorophenyl methan-1-ol, pubchem4664, acmc-209csd, intermediates-zcf02104, ksc494g9r
C1=C(C(=CC(=C1F)F)F)CO
162.111
162.11
≥98.0% (GC)

Chemical Identifiers

144284-25-3
162.111
NRXZCCOHXZFHBV-UHFFFAOYSA-N
2777035
C1=C(C(=CC(=C1F)F)F)CO
C7H5F3O
MFCD00061206
2,4,5-trifluorobenzyl alcohol, 2,4,5-trifluorophenyl methanol, 2,4,5-trifluorobenzylalcohol, benzenemethanol, 2,4,5-trifluoro, 2,4,5-trifluoro-phenyl-methanol, 2,4,5-trifluorophenyl methan-1-ol, pubchem4664, acmc-209csd, intermediates-zcf02104, ksc494g9r
(2,4,5-trifluorophenyl)methanol
Safety and Handling

Safety and Handling

TSCA : No

Documents