Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,5-Trifluorobenzyl alcohol, 98%

$169.32 - $169.32

Chemical Identifiers

Chemical Name or Material 2,4,5-Trifluorobenzyl alcohol
CAS 144284-25-3
Molecular Formula C7H5F3O
Formula Weight 162.11
MDL Number MFCD00061206
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1996006

 
alfa aesar™
A1996006
5g Each for $169.32
Description & Specifications

Specifications

Formula Weight 162.11
Chemical Name or Material 2,4,5-Trifluorobenzyl alcohol
Assay 98%
Molecular Formula C7H5F3O
MDL Number MFCD00061206
CAS 144284-25-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Flash Point 110°C (230°F)
Boiling Point 201° to 204°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.