missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,5-Trifluorobenzoyl chloride, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC309330050

 View more versions of this product

Catalog No. AC309330050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥98.5%
194.54
Clear colorless
100.0
F3C6H2COCl
C7H2ClF3O
99%
98.5
Glass bottle
Liquid
5g
1.5200g/mL
Authentic
2, 4, 5-Trifluorobenzoyl chloride
88419-56-1
MFCD00061204
1.4970 to 1.4990 (20°C, 589nm)

Chemical Identifiers

88419-56-1
100.0
C7H2ClF3O
194.54
2, 4, 5-Trifluorobenzoyl chloride
98.5
99%
F3C6H2COCl
MFCD00061204
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger