Please login to your online account to display your discounted pricing

2,4,5-Trifluorobenzoyl chloride, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 4, 5-Trifluorobenzoyl chloride
CAS 88419-56-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H2ClF3O
Linear Formula F3C6H2COCl
Formula Weight 194.54
MDL Number MFCD00061204
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC309330050

  SDS
thermo fisher scientific
AC309330050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC309330010

  SDS
thermo fisher scientific
309330010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 194.54
Quantity 5g
Color Clear colorless
Density 1.5200g/mL
CAS Max % 100.0
Infrared Spectrum Authentic
Linear Formula F3C6H2COCl
Chemical Name or Material 2, 4, 5-Trifluorobenzoyl chloride
Molecular Formula C7H2ClF3O
CAS 88419-56-1
Assay Percent Range 99%
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00061204
CAS Min % 98.5
Refractive Index 1.4970 to 1.4990 (20°C, 589nm)
Packaging Glass bottle

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.