Please login to your online account to display your discounted pricing

2,4,5-Trifluorobenzoyl chloride, 99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  309330010

 View more versions of this product

Catalog No. AC309330010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 194.54
Quantity 1g
Color Clear colorless
Density 1.5200g/mL
CAS Max % 100.0
Chemical Name or Material 2, 4, 5-Trifluorobenzoyl chloride
Packaging Glass bottle
Refractive Index 1.4970 to 1.4990 (20°C, 589nm)
Molecular Formula C7H2ClF3O
Linear Formula F3C6H2COCl
CAS 88419-56-1
Infrared Spectrum Authentic
Assay Percent Range 99%
MDL Number MFCD00061204
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2, 4, 5-Trifluorobenzoyl chloride
CAS 88419-56-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H2ClF3O
Linear Formula F3C6H2COCl
Formula Weight 194.54
MDL Number MFCD00061204
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents