Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,5-Trifluorobenzoyl chloride, 98%

Chemical Identifiers

Chemical Name or Material 2,4,5-Trifluorobenzoyl chloride
CAS 88419-56-1
Molecular Formula C7H2ClF3O
Formula Weight 194.54
MDL Number MFCD00061204
UN Number UN3265
Beilstein 3606259
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1860306

  SDS
alfa aesar™
A1860306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 194.54
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Chemical Name or Material 2,4,5-Trifluorobenzoyl chloride
UN Number UN3265
MDL Number MFCD00061204
Assay 98%
Quantity 5g
Beilstein 3606259
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Flash Point 87°C (188°F)
CAS 88419-56-1
Health Hazard 1 Danger
Molecular Formula C7H2ClF3O
Boiling Point 85°C (17mmHg)
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.