missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,5-Trifluorobenzonitrile, TCI America™

Manufacturer:  TCI America T232625G

 View more versions of this product

Catalog No. T232625G



Specifications

Specifications

2,4,5-Trifluorobenzonitrile
Yellow
98349-22-5
MFCD00013289
benzonitrile, 2,4,5-trifluoro, 2,4,5-trifluoro-benzonitrile, 2,4,5-trifluorobenzenecarbonitrile, 3-flurophenol, pubchem1571, acmc-209sa5, 2,4,5-trifluorophenylnitrile, ksc488c2t, 2,4,5-trifluoro benzonitrile, dlknogqoozficz-uhfffaoysa
C1=C(C(=CC(=C1F)F)F)C#N
157.095
157.10
≥98.0% (GC)
170°C
25g
C7H2F3N
3276
DLKNOGQOOZFICZ-UHFFFAOYSA-N
2,4,5-trifluorobenzonitrile
593813
Clear Liquid at 20°C

Chemical Identifiers

98349-22-5
157.095
DLKNOGQOOZFICZ-UHFFFAOYSA-N
593813
C1=C(C(=CC(=C1F)F)F)C#N
C7H2F3N
MFCD00013289
benzonitrile, 2,4,5-trifluoro, 2,4,5-trifluoro-benzonitrile, 2,4,5-trifluorobenzenecarbonitrile, 3-flurophenol, pubchem1571, acmc-209sa5, 2,4,5-trifluorophenylnitrile, ksc488c2t, 2,4,5-trifluoro benzonitrile, dlknogqoozficz-uhfffaoysa
2,4,5-trifluorobenzonitrile
Safety and Handling

Safety and Handling

TSCA : No

Documents