missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,4,5-Trifluorobenzenesulfonyl chloride, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC448880050
Specifications
2, 4, 5-Trifluorobenzenesulfonyl chloride | |
>100°C | |
1.657 | |
220227-21-4 | |
C6H2ClF3O2S | |
2,4,5-trifluoro-benzenesulfonyl chloride, 2,4,5-trifluorobenzene-1-sulfonyl chloride, 2,4,5-trifluorobenzenesulphonyl chloride, 2,4,5-trifluorophenyl sulfonyl chloride, 2,4,5-trifluorophenylsulfonyl chloride, acmc-209fqs, benzenesulfonyl chloride, 2,4,5-trifluoro, chloro 2,4,5-trifluorophenyl sulfone, ksc201o4t, pubchem5179 | |
C1=C(C(=CC(=C1F)S(=O)(=O)Cl)F)F | |
230.59 | |
230.59 |
1.6570g/mL | |
Glass bottle | |
5g | |
96% min. (GC) | |
F3C6H2SO2Cl | |
STCZWXXRRTWRSK-UHFFFAOYSA-N | |
2,4,5-trifluorobenzenesulfonyl chloride | |
2776979 | |
97% |
Chemical Identifiers
220227-21-4 | |
230.59 | |
2,4,5-trifluoro-benzenesulfonyl chloride, 2,4,5-trifluorobenzene-1-sulfonyl chloride, 2,4,5-trifluorobenzenesulphonyl chloride, 2,4,5-trifluorophenyl sulfonyl chloride, 2,4,5-trifluorophenylsulfonyl chloride, acmc-209fqs, benzenesulfonyl chloride, 2,4,5-trifluoro, chloro 2,4,5-trifluorophenyl sulfone, ksc201o4t, pubchem5179 | |
2,4,5-trifluorobenzenesulfonyl chloride |
C6H2ClF3O2S | |
STCZWXXRRTWRSK-UHFFFAOYSA-N | |
2776979 | |
C1=C(C(=CC(=C1F)S(=O)(=O)Cl)F)F |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Danger