Please login to your online account to display your discounted pricing

2,4,5-Trifluorobenzenesulfonyl chloride, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 2, 4, 5-Trifluorobenzenesulfonyl chloride
Assay Percent Range 97%
Molecular Formula C6H2ClF3O2S
Formula Weight 230.59
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC448880010

 
thermo fisher scientific
AC448880010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC448880050

 
thermo fisher scientific
AC448880050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 230.59
Quantity 1g
Chemical Name or Material 2, 4, 5-Trifluorobenzenesulfonyl chloride
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C6H2ClF3O2S
CAS 220227-21-4