Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,5-Trifluorobenzaldehyde, 97%

Chemical Identifiers

Chemical Name or Material 2,4,5-Trifluorobenzaldehyde
CAS 165047-24-5
Molecular Formula C7H3F3O
Formula Weight 160.1
MDL Number MFCD00061196
UN Number UN1989
Beilstein 8479631
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2098603

  SDS
alfa aesar™
B2098603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2098606

  SDS
alfa aesar™
B2098606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 160.1
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Molecular Formula C7H3F3O
CAS 165047-24-5
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, N.O.S.
Chemical Name or Material 2,4,5-Trifluorobenzaldehyde
MDL Number MFCD00061196
Boiling Point 174°C
UN Number UN1989
Quantity 1g
Health Hazard 1 Warning
Flash Point 58°C (136°F)
Assay 97%
Beilstein 8479631

Applications
It is a reactant in the enantioselective synthesis of (R)-selegiline, an anti-Parkinson′s drug, (S)-benzphetamine, an anti-obesity agent, and (S)-sitagliptin, an anti-diabetic drug.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.