Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',4',5'-Trifluoroacetophenone, 99%

$159.72 - $159.72

Chemical Identifiers

Chemical Name or Material 2',4',5'-Trifluoroacetophenone
CAS 129322-83-4
Molecular Formula C8H5F3O
Formula Weight 174.12
MDL Number MFCD00061193
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2620506

  SDS
alfa aesar™
H2620506
5g Each for $159.72
Description & Specifications

Specifications

Formula Weight 174.12
Assay 99%
Quantity 5g
Chemical Name or Material 2',4',5'-Trifluoroacetophenone
Molecular Formula C8H5F3O
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Flash Point 71°C (160°F)
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
CAS 129322-83-4
MDL Number MFCD00061193
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.