missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,5-Trifluoro-3-methoxybenzoic Acid, TCI America™

Manufacturer:  TCI America T191725G

 View more versions of this product

Catalog No. T191725G



Specifications

Specifications

2,4,5-Trifluoro-3-methoxybenzoic Acid
118°C
112811-65-1
MFCD00153201
YVJHZWWMKFQKDC-UHFFFAOYSA-N
2,4,5-trifluoro-3-methoxybenzoic acid
2733970
Crystal-Powder at 20°C
White
25g
C8H5F3O3
3-methoxy-2,4,5-trifluorobenzoic acid, 2,4,5-trifluoro-m-anisic acid, 2,4,5-trifluoro-3-methoxy benzoic acid, 2,4,5-trifluoro-3-methoxy-benzoic acid, benzoic acid, 2,4,5-trifluoro-3-methoxy, pubchem1352, pubchem2237, acmc-1bt5z, ksc181i7h, 3-methoxy-2,4,5-triflourobenzoic acid
COC1=C(C(=CC(=C1F)F)C(=O)O)F
206.12
206.12
≥98.0% (GC,T)

Chemical Identifiers

112811-65-1
206.12
YVJHZWWMKFQKDC-UHFFFAOYSA-N
2733970
COC1=C(C(=CC(=C1F)F)C(=O)O)F
C8H5F3O3
MFCD00153201
3-methoxy-2,4,5-trifluorobenzoic acid, 2,4,5-trifluoro-m-anisic acid, 2,4,5-trifluoro-3-methoxy benzoic acid, 2,4,5-trifluoro-3-methoxy-benzoic acid, benzoic acid, 2,4,5-trifluoro-3-methoxy, pubchem1352, pubchem2237, acmc-1bt5z, ksc181i7h, 3-methoxy-2,4,5-triflourobenzoic acid
2,4,5-trifluoro-3-methoxybenzoic acid
Safety and Handling

Safety and Handling

EINECSNumber : (3)-4378

TSCA : No

Documents