Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,5-Trifluoro-3-methoxybenzoic acid, 98%

$21.86 - $109.43

Chemical Identifiers

Chemical Name or Material 2,4,5-Trifluoro-3-methoxybenzoic acid
CAS 112811-65-1
Molecular Formula C8H5F3O3
Formula Weight 206.12
MDL Number MFCD00153201
Beilstein 7428474
Synonym 3-Methoxy-2,4,5-trifluorobenzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1923703

  SDS
alfa aesar™
L1923703
1g Each for $21.86

AAL1923706

  SDS
alfa aesar™
L1923706
5g Each for $109.43
Description & Specifications

Specifications

Formula Weight 206.12
Chemical Name or Material 2,4,5-Trifluoro-3-methoxybenzoic acid
Assay 98%
Molecular Formula C8H5F3O3
MDL Number MFCD00153201
CAS 112811-65-1
Melting Point 114° to 118°C
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 7428474
Synonym 3-Methoxy-2,4,5-trifluorobenzoic acid
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.