missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,5-Trifluoro-1-iodobenzene, 95%

Manufacturer:  Alfa Aesar™ H6147514

 View more versions of this product

Catalog No. AAH6147514



Specifications

Specifications

2,4,5-Trifluoro-1-iodobenzene
Pink
17533-08-3
MFCD04973898
UPCKPWDYXMTASO-UHFFFAOYSA-N
1,2,4-trifluoro-5-iodobenzene
2760713
Liquid
Light Sensitive
25g
C6H2F3I
1-iodo-2,4,5-trifluorobenzene
C1=C(C(=CC(=C1F)I)F)F
257.982
257.98
95%

Chemical Identifiers

17533-08-3
257.982
UPCKPWDYXMTASO-UHFFFAOYSA-N
2760713
C1=C(C(=CC(=C1F)I)F)F
C6H2F3I
MFCD04973898
1-iodo-2,4,5-trifluorobenzene
1,2,4-trifluoro-5-iodobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C

Documents