missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,5-Trichloropyrimidine, TCI America™

Manufacturer:  TCI America T251225G

 View more versions of this product

Catalog No. T251225G



Specifications

Specifications

2,4,5-Trichloropyrimidine
Yellow
5750-76-5
MFCD03788200
GIKMWFAAEIACRF-UHFFFAOYSA-N
2,4,5-trichloropyrimidine
237259
Clear Liquid at 20°C
96°C
25g
C4HCl3N2
2,5,6-trichloropyrimidine, 2,4,5-trichloro-pyrimidine, trichloropyrimidine, pyrimidine, 2,4,5-trichloro, 2,4,5-trichlorpyrimidin, pubchem5302, zlchem 1349, pyrimidine, trichloro, acmc-209lys, 2,5-trichloropyrimidine
C1=C(C(=NC(=N1)Cl)Cl)Cl
183.416
183.42
≥98.0% (GC)

Chemical Identifiers

5750-76-5
183.416
GIKMWFAAEIACRF-UHFFFAOYSA-N
237259
C1=C(C(=NC(=N1)Cl)Cl)Cl
C4HCl3N2
MFCD03788200
2,5,6-trichloropyrimidine, 2,4,5-trichloro-pyrimidine, trichloropyrimidine, pyrimidine, 2,4,5-trichloro, 2,4,5-trichlorpyrimidin, pubchem5302, zlchem 1349, pyrimidine, trichloro, acmc-209lys, 2,5-trichloropyrimidine
2,4,5-trichloropyrimidine
Safety and Handling

Safety and Handling

TSCA : No

Documents