missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,5-Trichlorophenyl isothiocyanate, 98%

Manufacturer:  Alfa Aesar™ L1155103

 View more versions of this product

Catalog No. AAL1155103



Specifications

Specifications

2,4,5-Trichlorophenyl isothiocyanate
141°C to 142°C (1mmHg)
1g
C7H2Cl3NS
UN2923
2,4,5-trichlorophenyl isothiocyanate, benzene, 1,2,4-trichloro-5-isothiocyanato, 2,4,5-trichlorophenylisothiocyanate, 2,4,5-trichlorophenyl isothiocyanate, acmc-1cb4v, 2,4,5-trichlorobenzenisothiocyanate, 1,2,4-trichloro-5-isothiocyanato-benzene, #, 2,4,5-trichlorophenyl isothiocyanate, 1,2,4-tris chloranyl-5-isothiocyanato-benzene
C1=C(C(=CC(=C1Cl)Cl)Cl)N=C=S
238.51
238.52
Irritating
42°C to 47°C
23165-46-0
MFCD00041052
881221
PJLRSYLEFZNICX-UHFFFAOYSA-N
1,2,4-trichloro-5-isothiocyanatobenzene
123408
98%

Chemical Identifiers

23165-46-0
238.51
PJLRSYLEFZNICX-UHFFFAOYSA-N
123408
C1=C(C(=CC(=C1Cl)Cl)Cl)N=C=S
C7H2Cl3NS
MFCD00041052
2,4,5-trichlorophenyl isothiocyanate, benzene, 1,2,4-trichloro-5-isothiocyanato, 2,4,5-trichlorophenylisothiocyanate, 2,4,5-trichlorophenyl isothiocyanate, acmc-1cb4v, 2,4,5-trichlorobenzenisothiocyanate, 1,2,4-trichloro-5-isothiocyanato-benzene, #, 2,4,5-trichlorophenyl isothiocyanate, 1,2,4-tris chloranyl-5-isothiocyanato-benzene
1,2,4-trichloro-5-isothiocyanatobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

Documents