missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,5-Trichlorobenzenesulfonyl chloride 97%, ACROS Organics™

Chemical Identifiers

CAS 15945-07-0
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C6 H2 Cl4 O2 S
Linear Formula Cl3C6H2SO2Cl
Formula Weight 279.94
MDL Number MFCD00007428
Chemical Name or Material 2, 4, 5-Trichlorobenzenesulfonyl chloride
Assay ≥96.0% (GC)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC139410050 SDS Thermo Fisher Scientific
AC139410050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC139410250 SDS Thermo Fisher Scientific
AC139410250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

15945-07-0
100.0
C6 H2 Cl4 O2 S
279.94
2, 4, 5-Trichlorobenzenesulfonyl chloride
96.0
97%
Cl3C6H2SO2Cl
MFCD00007428
≥96.0% (GC)

Specifications

5g
96.0
Liquid
C6 H2 Cl4 O2 S
Authentic
MFCD00007428
66° to 70°C
2, 4, 5-Trichlorobenzenesulfonyl chloride
15945-07-0
100.0
97%
Cl3C6H2SO2Cl
279.94
Glass bottle
Clear yellow to orange to slightly brown, may darken on storage
≥96.0% (GC)

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger