Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,5-Trichlorobenzenesulfonic acid potassium salt, 90+%

$33.76 - $506.27

Chemical Identifiers

Chemical Name or Material 2,4,5-Trichlorobenzenesulfonic acid potassium salt
CAS 62625-17-6
Molecular Formula C6H2Cl3KO3S
Formula Weight 299.6
MDL Number MFCD00024894
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2071314

  SDS
alfa aesar™
B2071314
25g Each for $506.27

AAB2071303

  SDS
alfa aesar™
B2071303
1g Each for $33.76
Description & Specifications

Specifications

Formula Weight 299.6
Chemical Name or Material 2,4,5-Trichlorobenzenesulfonic acid potassium salt
TSCA TSCA
Assay ≥90%
Molecular Formula C6H2Cl3KO3S
MDL Number MFCD00024894
CAS 62625-17-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point >300°C
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.