missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,5-Trichlorobenzenesulfonic Acid Hydrate, TCI America™

$120.70 - $363.63

Chemical Identifiers

CAS 6378-25-2
Molecular Formula C6H3Cl3O3S
Molecular Weight (g/mol) 261.497
MDL Number MFCD00070539
InChI Key LEDKKDPOPIKMSZ-UHFFFAOYSA-N
PubChem CID 80786
IUPAC Name 2,4,5-trichlorobenzenesulfonic acid
SMILES C1=C(C(=CC(=C1Cl)Cl)Cl)S(=O)(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B02485G View Documents TCI America
B02485G
5g Each for $120.70
 
B024825G View Documents TCI America
B024825G
25g Each for $363.63
 
Specifications

Chemical Identifiers

6378-25-2
261.497
LEDKKDPOPIKMSZ-UHFFFAOYSA-N
2,4,5-trichlorobenzenesulfonic acid
C6H3Cl3O3S
MFCD00070539
80786
C1=C(C(=CC(=C1Cl)Cl)Cl)S(=O)(=O)O

Specifications

White
6378-25-2
MFCD00070539
LEDKKDPOPIKMSZ-UHFFFAOYSA-N
2,4,5-trichlorobenzenesulfonic acid
80786
Crystal-Powder at 20°C
2,4,5-Trichlorobenzenesulfonic Acid Hydrate
5g
C6H3Cl3O3S
2585
C1=C(C(=CC(=C1Cl)Cl)Cl)S(=O)(=O)O
261.497
261.50
≥98.0% (T)
Safety and Handling

Safety and Handling

TSCA : No