Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,5-Trichlorobenzenesulfonamide, 97%

$117.30 - $375.58

Chemical Identifiers

Chemical Name or Material 2,4,5-Trichlorobenzenesulfonamide
CAS 29092-27-1
Molecular Formula C6H4Cl3NO2S
Formula Weight 260.53
MDL Number MFCD00119361
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2768703

  SDS
alfa aesar™
H2768703
1g Each for $117.30

AAH2768706

  SDS
alfa aesar™
H2768706
5g Each for $375.58
Description & Specifications

Specifications

Formula Weight 260.53
Chemical Name or Material 2,4,5-Trichlorobenzenesulfonamide
Assay 97%
Molecular Formula C6H4Cl3NO2S
MDL Number MFCD00119361
CAS 29092-27-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 187° to 191°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.