Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,5-Tribromotoluene, 97+%

$47.32 - $779.00

Chemical Identifiers

Chemical Name or Material 2,4,5-Tribromotoluene
CAS 3278-88-4
Molecular Formula C7H5Br3
Formula Weight 328.83
MDL Number MFCD00052984
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1350703

  SDS
alfa aesar™
A1350703
1g Each for $47.32

AAA1350706

  SDS
alfa aesar™
A1350706
5g Each for $180.81

AAA1350714

  SDS
alfa aesar™
A1350714
25g Each for $779.00
Description & Specifications

Specifications

Formula Weight 328.83
Chemical Name or Material 2,4,5-Tribromotoluene
Assay ≥97%
Molecular Formula C7H5Br3
MDL Number MFCD00052984
CAS 3278-88-4
Melting Point 112° to 115°C
Health Hazard 1 Warning
Quantity 1g
Odor Odorless
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.