Please login to your online account to display your discounted pricing

2,4,5-Tribromoimidazole, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  161640250

 View more versions of this product

Catalog No. AC161640250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 304.76
Physical Form Powder
Quantity 25g
Color Beige-yellow
Beilstein 23, 49
CAS Max % 100.0
CAS Min % 96.0
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00005184
Chemical Name or Material 2, 4, 5-Tribromoimidazole
CAS 2034-22-2
Assay Percent Range 97%
Molecular Formula C3HBr3N2
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material 2, 4, 5-Tribromoimidazole
CAS 2034-22-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C3HBr3N2
Formula Weight 304.76
MDL Number MFCD00005184
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.

SDS & Safety Documents