Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,5-Tribromoimidazole, 97%

Chemical Identifiers

Chemical Name or Material 2,4,5-Tribromoimidazole
CAS 2034-22-2
Molecular Formula C3HBr3N2
Formula Weight 304.78
MDL Number MFCD00005184
UN Number UN2811
Beilstein 115314
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0571606

  SDS
alfa aesar™
L0571606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0571614

  SDS
alfa aesar™
L0571614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 304.78
Chemical Name or Material 2,4,5-Tribromoimidazole
Molecular Formula C3HBr3N2
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 2034-22-2
UN Number UN2811
Health Hazard 2 GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Odor Odorless
RTECS Number NI8660000
MDL Number MFCD00005184
Quantity 5g
Melting Point ≈210°C
Beilstein 115314
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.