Please login to your online account to display your discounted pricing

2,4,5,6-Tetrachloropyrimidine, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2,4,5,6-Tetrachloropyrimidine
CAS 1780-40-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C4Cl4N2
Formula Weight 217.87
MDL Number MFCD00006062
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC333520010

  SDS
thermo fisher scientific
AC333520010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥96.0% (GC)
Formula Weight 217.87
Quantity 1g
Color White
RTECS Number UV9745000
EINECS Number 217-228-3
TSCA TSCA
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 2,4,5,6-Tetrachloropyrimidine
Infrared Spectrum Authentic
Assay Percent Range 97%
Health Hazard 1 Warning
Health Hazard 2 GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Molecular Formula C4Cl4N2
MDL Number MFCD00006062
CAS 1780-40-1
Safety and Handling

Warning

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.