missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,4-Trimethyl-2-pentanol, TCI America™

Manufacturer:  TCI America T19621G

 View more versions of this product

Catalog No. T19621G



Specifications

Specifications

2,4,4-Trimethyl-2-pentanol
1g
C8H18O
1987
BSYJHYLAMMJNRC-UHFFFAOYSA-N
2,4,4-trimethylpentan-2-ol
94740
Clear Liquid at 20°C
Yellow
690-37-9
MFCD00101611
2-Hydroxy-2,4,4-trimethylpentane
CC(C)(C)CC(C)(C)O
130.231
130.23
≥95.0% (GC)

Chemical Identifiers

690-37-9
130.231
BSYJHYLAMMJNRC-UHFFFAOYSA-N
94740
CC(C)(C)CC(C)(C)O
C8H18O
MFCD00101611
2-Hydroxy-2,4,4-trimethylpentane
2,4,4-trimethylpentan-2-ol
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0217

TSCA : No

Documents