missing translation for 'onlineSavingsMsg'
Learn More

2,4,4-Trimethyl-1-pentene 98.0+%, TCI America™

Supplier:  TCI America T0665100ML

 View more versions of this product

Catalog No. T0665100ML

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,4,4-Trimethyl-1-pentene
-93°C
100°C
MFCD00008855
2050
FXNDIJDIPNCZQJ-UHFFFAOYSA-N
2,4,4-trimethylpent-1-ene
7868
≥98.0% (GC)
107-39-1
Colorless
C8H16
100 mL
2,4,4-trimethyl-1-pentene, diisobutylene, pentene, 2,4,4-trimethyl, 2,4,4-trimethylpentene, 1-pentene, 2,4,4-trimethyl, 2,2,4-trimethyl-4-pentene, unii-n69l73advf, 1-methyl-1-neopentylethylene, ccris 9103, 2,4,4-trimethylpentene-1
CC(=C)CC(C)(C)C
112.216
112.22
Liquid

Chemical Identifiers

107-39-1
112.216
FXNDIJDIPNCZQJ-UHFFFAOYSA-N
7868
CC(=C)CC(C)(C)C
C8H16
MFCD00008855
2,4,4-trimethyl-1-pentene, diisobutylene, pentene, 2,4,4-trimethyl, 2,4,4-trimethylpentene, 1-pentene, 2,4,4-trimethyl, 2,2,4-trimethyl-4-pentene, unii-n69l73advf, 1-methyl-1-neopentylethylene, ccris 9103, 2,4,4-trimethylpentene-1
2,4,4-trimethylpent-1-ene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0024

RTECSNumber : SB2715000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions