Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,4'-Trimethoxybenzophenone, 99%

$69.46 - $244.09

Chemical Identifiers

Chemical Name or Material 2,4,4'-Trimethoxybenzophenone
CAS 4038-15-7
Molecular Formula C16H16O4
Formula Weight 272.3
MDL Number MFCD00100089
Beilstein 1991889
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1358009

  SDS
alfa aesar™
L1358009
10g Each for $69.46

AAL1358018

  SDS
alfa aesar™
L1358018
50g Each for $244.09
Description & Specifications

Specifications

Formula Weight 272.3
Chemical Name or Material 2,4,4'-Trimethoxybenzophenone
MDL Number MFCD00100089
Molecular Formula C16H16O4
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 99%
Beilstein 1991889
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 71° to 73°C
Quantity 10g
CAS 4038-15-7
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.