Please login to your online account to display your discounted pricing

2,4,4'-Trihydroxybenzophenone, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  139810050

 View more versions of this product

Catalog No. AC139810050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Powder or Chunks
Formula Weight 230.22
Quantity 5g
Color Orange
Beilstein 08, 422
TSCA TSCA
EINECS Number 216-004-2
CAS Min % 94.0
CAS Max % 100.0
MDL Number MFCD00002356
Infrared Spectrum Authentic
Chemical Name or Material 2, 4, 4'-Trihydroxybenzophenone
Molecular Formula C13H10O4
Linear Formula (HO)2C6H3COC6H4OH
CAS 1470-79-7

Specifications

Chemical Name or Material 2, 4, 4'-Trihydroxybenzophenone
CAS 1470-79-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Molecular Formula C13H10O4
Linear Formula (HO)2C6H3COC6H4OH
Formula Weight 230.22
MDL Number MFCD00002356