Please login to your online account to display your discounted pricing

2,4,4'-Trihydroxybenzophenone, ACROS Organics™

$214.73 - $214.73

Chemical Identifiers

Chemical Name or Material 2, 4, 4'-Trihydroxybenzophenone
CAS 1470-79-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Molecular Formula C13H10O4
Linear Formula (HO)2C6H3COC6H4OH
Formula Weight 230.22
MDL Number MFCD00002356
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC139810050

  SDS
thermo fisher scientific
139810050
5g Each for $214.73
Description & Specifications

Specifications

Assay ≥94%
Physical Form Powder or Chunks
Formula Weight 230.22
Quantity 5g
Color Orange
Beilstein 08, 422
TSCA TSCA
EINECS Number 216-004-2
CAS Min % 94.0
CAS Max % 100.0
MDL Number MFCD00002356
Infrared Spectrum Authentic
Chemical Name or Material 2, 4, 4'-Trihydroxybenzophenone
Molecular Formula C13H10O4
Linear Formula (HO)2C6H3COC6H4OH
CAS 1470-79-7