Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4,4,6-Tetramethyl-1-oxa-3-aza-2-cyclohexene, 95%

Chemical Identifiers

Chemical Name or Material 2,4,4,6-Tetramethyl-1-oxa-3-aza-2-cyclohexene
CAS 26939-18-4
Molecular Formula C8H15NO
Formula Weight 141.22
MDL Number MFCD00014605
UN Number UN1993
Beilstein 111529
Synonym 2,4,4,6-Tetramethyl-5,6-dihydro-1,3(4H)-oxazine; 5,6-Dihydro-2,4,4,6-tetramethyl-1,3(4H)-oxazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2043806

  SDS
alfa aesar™
B2043806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2043814

  SDS
alfa aesar™
B2043814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 141.22
Boiling Point 147° to 149°C
Quantity 5g
Chemical Name or Material 2,4,4,6-Tetramethyl-1-oxa-3-aza-2-cyclohexene
MDL Number MFCD00014605
CAS 26939-18-4
Assay 95%
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
TSCA TSCA
UN Number UN1993
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 111529
Synonym 2,4,4,6-Tetramethyl-5,6-dihydro-1,3(4H)-oxazine; 5,6-Dihydro-2,4,4,6-tetramethyl-1,3(4H)-oxazine
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Molecular Formula C8H15NO
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapour.