missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,4,6-Tetrabromo-2,5-cyclohexadien-1-one, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 257690050

 View more versions of this product

Catalog No. AC257690050



Description

Description

C-X bond formation reagent
Specifications

Specifications

2,4,4,6-Tetrabromo-2,5-cyclohexadien-1-one
Authentic
Colorless
5g
20244-61-5
100.0
C6H2Br4O
12,457; 16,316
NJQJGRGGIUNVAB-UHFFFAOYSA-N
2,4,4,6-tetrabromocyclohexa-2,5-dien-1-one
88433
Liquid
94% min.
Glass bottle
120°C to 127°C
95%
94.0
94% min. (Iodometry)
MFCD00001589
2,2,4,4-tetrabromo-2,5-cyclohexadienone, 2,4,4,6-tetrabromo-2,5-cyclohexadien-1-one, 2,4,4,6-tetrabromo-2,5-cyclohexadienone, 2,4,4,6-tetrabromocyclohexa-2,5-dienone, 2,4,4,6-tetrabromocyclohexadiene-1-one, 2,4,4,6-tetrabromocyclohexadienone, 2,5-cyclohexadien-1-one, 2,4,4,6-tetrabromo, acmc-1cce4, ksc496q7n, tbco
C1=C(C(=O)C(=CC1(Br)Br)Br)Br
409.69
409.69
95%

Chemical Identifiers

20244-61-5
409.69
NJQJGRGGIUNVAB-UHFFFAOYSA-N
88433
C1=C(C(=O)C(=CC1(Br)Br)Br)Br
C6H2Br4O
MFCD00001589
2,2,4,4-tetrabromo-2,5-cyclohexadienone, 2,4,4,6-tetrabromo-2,5-cyclohexadien-1-one, 2,4,4,6-tetrabromo-2,5-cyclohexadienone, 2,4,4,6-tetrabromocyclohexa-2,5-dienone, 2,4,4,6-tetrabromocyclohexadiene-1-one, 2,4,4,6-tetrabromocyclohexadienone, 2,5-cyclohexadien-1-one, 2,4,4,6-tetrabromo, acmc-1cce4, ksc496q7n, tbco
2,4,4,6-tetrabromocyclohexa-2,5-dien-1-one
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 243-638-7

TSCA : TSCA

SDS Documents