Please login to your online account to display your discounted pricing

2,4,4,6-Tetrabromo-2,5-cyclohexadien-1-one, 95%, ACROS Organics™

$71.22 - $71.22

Chemical Identifiers

Chemical Name or Material 2,4,4,6-Tetrabromo-2,5-cyclohexadien-1-one
Name Note 95%
CAS 20244-61-5
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94% (Iodometry)
Assay Percent Range 95%
Molecular Formula C6H2Br4O
Formula Weight 409.69
MDL Number MFCD00001589
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC257690050

  SDS
thermo fisher scientific
257690050
Glass bottle 5g Each for $71.22

AC257690250

  SDS
fisher scientific acros
257690250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94% (Iodometry)
Physical Form Liquid
Formula Weight 409.69
Quantity 5g
Color Clear colorless
EINECS Number 243-638-7
TSCA TSCA
Fieser 12,457; 16,316
CAS Min % 94.0
CAS Max % 100.0
Elemental Analysis 94% min.
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2,4,4,6-Tetrabromo-2,5-cyclohexadien-1-one
Name Note 95%
Assay Percent Range 95%
MDL Number MFCD00001589
Molecular Formula C6H2Br4O
CAS 20244-61-5

C-X bond formation reagent

Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.