Please login to your online account to display your discounted pricing

2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Name Note 97%
CAS 269409-97-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C12H17BO3
Formula Weight 220.08
MDL Number MFCD02093754
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC366400050

  SDS
thermo fisher scientific
AC366400050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 220.08
Physical Form Liquid
Quantity 5g
Color Pale yellow to brown
Density 1.05g/mL
CAS Min % 96.0
CAS Max % 100.0
MDL Number MFCD02093754
Assay Percent Range 97%
Recommended Storage Refrigerator +4°C
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Molecular Formula C12H17BO3
Infrared Spectrum Authentic
Refractive Index 1.5060 to 1.5080 (20°C, 589nm)
Packaging Glass bottle
Name Note 97%
Chemical Name or Material 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
CAS 269409-97-4
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.