Please login to your online account to display your discounted pricing

2'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)furan 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)furan
CAS 374790-93-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H15BO3
Formula Weight 194.04
MDL Number MFCD02094008
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC390850050

  SDS
thermo fisher scientific
AC390850050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 194.04
Quantity 5g
Color Clear colorless to pale yellow
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 2'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)furan
Assay Percent Range 97%
Molecular Formula C10H15BO3
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD02094008
Packaging Glass bottle
Refractive Index 1.4730 to 1.4750 (20°C, 589nm)
CAS 374790-93-9
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.