Learn More
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 367560010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline | |
191171-55-8 | |
100.0 | |
96% min. (GC) | |
C12H18BNO2 | |
1g | |
Solubility in water: insoluble in water. | |
CC1(C)OB(OC1(C)C)C1=CC=CC=C1N | |
219.09 | |
219.09 |
97% | |
96.0 | |
Authentic | |
Glass bottle | |
MFCD02179448 | |
2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 2-aminophenylboronic acid pinacol ester, 2-aminophenylboronic acid, pinacol ester, 2-aminophenylboronicacidpinacolester, 2-aminophenylboronic acid pinacol cyclic ester, 2-aminobenzeneboronic acid pinacol ester, 2-aminophenyl boronic acid pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzenamine, 2-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, benzenamine, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
ZCJRWQDZPIIYLM-UHFFFAOYSA-N | |
2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline | |
2734652 | |
97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
Recommended Storage : product may turn dark on exposure to air
RUO â Research Use Only