Learn More
2-[3-(Trifluoromethyl)phenyl]ethylamine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6408403
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-[3-(Trifluoromethyl)phenyl]ethylamine | |
Colorless to Yellow | |
113°C (22 mmHg) | |
1.4645 | |
1 g | |
4398117 | |
2-3-trifluoromethyl phenyl ethanamine, 3-trifluoromethyl phenethylamine, 2-3-trifluoromethylphenyl ethylamine, 2-3-trifluoromethyl phenyl ethylamine, phenethylamine, m-trifluoromethyl, m-trifluoromethylphenethylamine, m-trifluoromethyl phenethylamine, benzeneethanamine, 3-trifluoromethyl, 2-3-trifluoromethylphenyl-ethylamine | |
C1=CC(=CC(=C1)C(F)(F)F)CCN | |
189.181 | |
189.18 | |
Liquid |
52516-30-0 | |
1.18 g/mL | |
C9H10F3N | |
MFCD00040756 | |
UN2735 | |
Light sensitive | |
BPVYCXMGJPKOTQ-UHFFFAOYSA-N | |
2-[3-(trifluoromethyl)phenyl]ethanamine | |
104223 | |
98% |
Chemical Identifiers
52516-30-0 | |
189.181 | |
BPVYCXMGJPKOTQ-UHFFFAOYSA-N | |
104223 | |
C1=CC(=CC(=C1)C(F)(F)F)CCN |
C9H10F3N | |
MFCD00040756 | |
2-3-trifluoromethyl phenyl ethanamine, 3-trifluoromethyl phenethylamine, 2-3-trifluoromethylphenyl ethylamine, 2-3-trifluoromethyl phenyl ethylamine, phenethylamine, m-trifluoromethyl, m-trifluoromethylphenethylamine, m-trifluoromethyl phenethylamine, benzeneethanamine, 3-trifluoromethyl, 2-3-trifluoromethylphenyl-ethylamine | |
2-[3-(trifluoromethyl)phenyl]ethanamine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber : 257-988-3
RTECSNumber : SI2525000
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only