missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-[3-(Trifluoromethyl)phenyl]acetamidoxime, 97%, Alfa Aesar™

Chemical Identifiers

CAS 925393-51-7
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66304MD View Documents Thermo Fisher Scientific
H66304MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6630403 View Documents Thermo Fisher Scientific
H6630403
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

925393-51-7

Specifications

White
925393-51-7
Solid
2-[3-(Trifluoromethyl)phenyl]acetamidoxime
250mg
218.18
97%
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures