Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-[3-(Trifluoromethyl)phenoxy]acetamidoxime, 97%

$373.78 - $373.78

Chemical Identifiers

Chemical Name or Material 2-[3-(Trifluoromethyl)phenoxy]acetamidoxime
CAS 690632-15-6
Molecular Formula C9H9F3N2O2
Formula Weight 234.18g/mol
MDL Number MFCD08060955
Synonym N-Hydroxy-2-[3-(trifluoromethyl)phenoxy]acetamidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5066306

 
alfa aesar™
H50663-06
Each for $373.78
Description & Specifications

Specifications

Quantity 5g
Chemical Name or Material 2-[3-(Trifluoromethyl)phenoxy]acetamidoxime
CAS 690632-15-6
Assay 97%
Molecular Formula C9H9F3N2O2
Formula Weight 234.18g/mol
MDL Number MFCD08060955
Synonym N-Hydroxy-2-[3-(trifluoromethyl)phenoxy]acetamidine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.