Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(3-Trifluoromethyl-alpha-toluenesulfonyl)thioacetamide, 97%

$235.27 - $235.27

Chemical Identifiers

Chemical Name or Material 2-(3-Trifluoromethyl-alpha-toluenesulfonyl)thioacetamide
CAS 175276-83-2
Molecular Formula C10H10F3NO2S2
Formula Weight 297.32g/mol
MDL Number MFCD00052940
Synonym 2-[3-(Trifluoromethyl)benzylsulfonyl]thioacetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2626306

 
alfa aesar™
H26263-06
5g Each for $235.27
Description & Specifications

Specifications

Melting Point 165° to 168°C
Quantity 5g
Chemical Name or Material 2-(3-Trifluoromethyl-alpha-toluenesulfonyl)thioacetamide
CAS 175276-83-2
Assay 97%
Molecular Formula C10H10F3NO2S2
Formula Weight 297.32g/mol
MDL Number MFCD00052940
Synonym 2-[3-(Trifluoromethyl)benzylsulfonyl]thioacetamide
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.